2,6-Dichlor-p-benzochinon 95%

Lieferant: Apollo Scientific

Synonyme: 2,6-Dichloro-1,4-benzoquinone

OR59912-5G
APOSOR59912-5GEA 108 EUR
APOSOR59912-5G
2,6-Dichlor-p-benzochinon 95%
2,6-Dichlor-p-benzochinon
Formel: C₆H₂Cl₂O₂
Molecular Weight: 176.99 g/mol
Schmelzpunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00037159
CAS-Nummer: 697-91-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR