Diethyl ethylidenemalonate 95+%

Lieferant: Apollo Scientific

OR59915-10G OR59915-25G
APOSOR59915-10GEA 77 EUR
APOSOR59915-10G APOSOR59915-25G
Diethyl ethylidenemalonate 95+%
Diethyl ethylidenemalonate
Formel: C₉H₁₄O₄
Siedepunkt: 115…118 °C/17mmHg
Dichte: 1,019 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009145
CAS-Nummer: 1462-12-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR