5-Chlor-2-iodpyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR59920-5G
OR59920-1G
OR59920-10G
APOSOR59920-5GEA
29
EUR
APOSOR59920-5G
APOSOR59920-1G
APOSOR59920-10G
5-Chlor-2-iodpyridin
5-Chlor-2-iodpyridin
Formel:
C₅H₃ClIN Schmelzpunkt: 85…87 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07368864 CAS-Nummer: 244221-57-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...