4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Formylphenyl)-5-methyl-1,3,4-oxadiazole

OR59953-250MG
APOSOR59953-250MGEA 58 EUR
APOSOR59953-250MG
4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzaldehyde 95%
4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzaldehyde
Formel: C₁₀H₈N₂O₂
Molecular Weight: 188.183 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11109323
CAS-Nummer: 179056-82-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR