5-Piperidino-2-thiophenecarbaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Piperidin-1-yl)thiophene-2-carbaldehyde

OR59954-1G
APOSOR59954-1GEA 139 EUR
APOSOR59954-1G
5-Piperidino-2-thiophenecarbaldehyde 97%
5-Piperidino-2-thiophenecarbaldehyde
Formel: C₁₀H₁₃NOS
Molecular Weight: 195.29 g/mol
Schmelzpunkt: 94…96 °C
Lagertemperatur: Kühlschrank
MDL: MFCD05263177
CAS-Nummer: 24372-48-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR