4-(4-Cyanphenyl)morpholin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Morpholinobenzonitrile, 4-(4-Morpholinyl)benzonitrile

OR59955-1G
APOSOR59955-1GEA 21.9 EUR
APOSOR59955-1G
4-(4-Cyanphenyl)morpholin 97%
4-(4-Cyanphenyl)morpholin
Formel: C₁₁H₁₂N₂O
Molecular Weight: 188,23 g/mol
Schmelzpunkt: 85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00100115
CAS-Nummer: 10282-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR