Quinoxaline-2-carbonyl chloride

Lieferant: Apollo Scientific

OR59969-1G
APOSOR59969-1GEA 171 EUR
APOSOR59969-1G
Quinoxaline-2-carbonyl chloride
Quinoxaline-2-carbonyl chloride
Formel: C₉H₅ClN₂O
Schmelzpunkt: 113…115 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00006725
CAS-Nummer: 54745-92-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR