Methyl-2-bromo-1-methyl-1H-imidazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

OR59983-250MG OR59983-1G
APOSOR59983-250MGEA 132 EUR
APOSOR59983-250MG APOSOR59983-1G
Methyl-2-bromo-1-methyl-1H-imidazole-5-carboxylate
Methyl-2-bromo-1-methyl-1H-imidazole-5-carboxylate
Formel: C₆H₇BrN₂O₂
Schmelzpunkt: 70…72 °C
MDL: MFCD02179563
CAS-Nummer: 120781-02-4
UN: 2811
ADR: 6,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR