1,5-Diaminopentan 95%
Lieferant: Apollo Scientific
OR59990-10G
APOSOR59990-10GEA
190
EUR
APOSOR59990-10G
1,5-Diaminopentan 95%
1,5-Diaminopentan
Formel:
C₅H₁₄N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008239 CAS-Nummer: 462-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...