N-Methyl-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)methylamine 97%

Lieferant: Apollo Scientific

Synonyme: 5-Methyl-4-[methyl(aminomethyl)]-1-phenyl-1H-pyrazole

OR59992-1G
APOSOR59992-1GEA 371 EUR
APOSOR59992-1G
N-Methyl-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)methylamine 97%
N-Methyl-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)methylamine
Formel: C₁₂H₁₅N₃
Molecular Weight: 201,268 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109316
CAS-Nummer: 1031843-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR