1-(4-Chlorophenylsulphonyl)-3,3-dimethyl-2-butanone 98%

Lieferant: Apollo Scientific
Achtung

OR6000-2.5G
APOSOR6000-2.5GEA 0 EUR
APOSOR6000-2.5G
1-(4-Chlorophenylsulphonyl)-3,3-dimethyl-2-butanone 98%
1-(4-Chlorophenylsulphonyl)-3,3-dimethyl-2-butanone
Formel: C₁₂H₁₅ClO₃S
Molecular Weight: 274,77 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082685
CAS-Nummer: 207974-06-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR