1-(2-Tetrahydropyranyl)-1H-pyrazol-5-boronsäurepinakolester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(Tetrahydropyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole-5-boronic acid Pinacol ester, 1-(Tetrahydropyran-2-yl)-1H-pyrazole-5-boronic acid pinacol ester, 1-(Tetrahydro-2H-pyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
OR60006-100G
APOSOR60006-100GEA
75.4
EUR
APOSOR60006-100G
1-(2-Tetrahydropyranyl)-1H-pyrazol-5-boronsäurepinakolester
1-(2-Tetrahydropyranyl)-1H-pyrazol-5-boronsäurepinakolester
Formel:
C₁₄H₂₃BN₂O₃ Molecular Weight: 278.16 g/mol Schmelzpunkt: 74…78 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09037501 CAS-Nummer: 903550-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...