1,3-Oxazole-4-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Cyano-1,3-oxazole, 4-Oxazolecarbonitrile

OR60008-1G
APOSOR60008-1GEA 102 EUR
APOSOR60008-1G
1,3-Oxazole-4-carbonitrile
1,3-Oxazole-4-carbonitrile
Formel: C₄H₂N₂O
Lagertemperatur: Kühlschrank
MDL: MFCD08059302
CAS-Nummer: 55242-84-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR