4-Indolboronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

OR60010-1G
APOSOR60010-1GEA 21.9 EUR
APOSOR60010-1G
4-Indolboronsäurepinakolester
4-Indolboronsäurepinakolester
Formel: C₁₄H₁₈BNO₂
Molecular Weight: 243.11 g/mol
Schmelzpunkt: 168…173 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689896
CAS-Nummer: 388116-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR