tert-Butyl (5-bromopyrazin-2-yl)carbamate 98%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-5-bromopyrazine, 2-BOC protected

OR60011-1G
APOSOR60011-1GEA 85.8 EUR
APOSOR60011-1G
tert-Butyl (5-bromopyrazin-2-yl)carbamate 98%
tert-Butyl (5-bromopyrazin-2-yl)carbamate
Formel: C₉H₁₂BrN₃O₂
Molecular Weight: 274,1100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08275683
CAS-Nummer: 914349-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR