2-Chloropyrimidine-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR60023-1G OR60023-250MG
APOSOR60023-1GEA 0 EUR
APOSOR60023-1G APOSOR60023-250MG
2-Chloropyrimidine-4-carboxylic acid
2-Chloropyrimidine-4-carboxylic acid
Formel: C₅H₃ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06739041
CAS-Nummer: 149849-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR