3-Aminopropylphosphonic acid

Lieferant: Apollo Scientific
Achtung

OR60033-1G OR60033-5G
APOSOR60033-1GEA 65.7 EUR
APOSOR60033-1G APOSOR60033-5G
3-Aminopropylphosphonic acid
3-Aminopropylphosphonic acid
Formel: C3H10NO3P
Molecular Weight: 139.0902 g/mol
Schmelzpunkt: 294 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008222
CAS-Nummer: 13138-33-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR