3-Aminopropylphosphonic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR60033-1G
OR60033-5G
APOSOR60033-1GEA
65.7
EUR
APOSOR60033-1G
APOSOR60033-5G
3-Aminopropylphosphonic acid
3-Aminopropylphosphonic acid
Formel:
C3H10NO3P Molecular Weight: 139.0902 g/mol Schmelzpunkt: 294 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008222 CAS-Nummer: 13138-33-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...