3,4-Dimethylbenzylamin 95%

Lieferant: Apollo Scientific
Gefahr

OR60034-5G
APOSOR60034-5GEA 55.1 EUR
APOSOR60034-5G
3,4-Dimethylbenzylamin 95%
3,4-Dimethylbenzylamin
Formel: C₉H₁₃N
Molecular Weight: 135.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00025577
CAS-Nummer: 102-48-7
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR