4-Iodbenzylalkohol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Iodophenyl)methanol
OR60044-5G
OR60044-10G
OR60044-25G
APOSOR60044-5GEA
26.1
EUR
APOSOR60044-5G
APOSOR60044-10G
APOSOR60044-25G
4-Iodbenzylalkohol
4-Iodbenzylalkohol
Formel:
C₇H₇IO Molecular Weight: 234.04 g/mol Schmelzpunkt: 72…75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01732720 CAS-Nummer: 18282-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...