2-Amino-2-(3-bromphenyl)essigsäure 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-DL-phenylglycine
OR60052-250MG
APOSOR60052-250MGEA
21.9
EUR
APOSOR60052-250MG
2-Amino-2-(3-bromphenyl)essigsäure 95+%
2-Amino-2-(3-bromphenyl)essigsäure
Formel:
C₈H₈BrNO₂ Molecular Weight: 230,0600 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00665358 CAS-Nummer: 79422-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...