Ethyl 2-bromo-5-chloro-1,3-thiazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-5-chloro-4-(ethoxycarbonyl)-1,3-thiazole, Ethyl 2-bromo-5-chlorothiazole-4-carboxylate

OR60060-1G
APOSOR60060-1GEA 65.4 EUR
APOSOR60060-1G
Ethyl 2-bromo-5-chloro-1,3-thiazole-4-carboxylate
Ethyl 2-bromo-5-chloro-1,3-thiazole-4-carboxylate
Formel: C₆H₅BrClNO₂S
Molecular Weight: 270.531 g/mol
Schmelzpunkt: 56 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08704625
CAS-Nummer: 425392-44-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR