1H-Indazol-6-carbonsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR60063-1G
APOSOR60063-1GEA
21.9
EUR
APOSOR60063-1G
1H-Indazol-6-carbonsäure 95%
1H-Indazole-6-carboxylic acid
Formel:
C₈H₆N₂O₂ Molecular Weight: 162,1454 g/mol Schmelzpunkt: 297…300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06804571 CAS-Nummer: 704-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...