4-Isochinolincarbonsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Isoquinoline-4-carboxylic acid
OR60073-5G
APOSOR60073-5GEA
190
EUR
APOSOR60073-5G
4-Isochinolincarbonsäure 98%
4-Isoquinolinecarboxylic acid
Formel:
C₁₀H₇NO₂ Molecular Weight: 173.1681 g/mol Schmelzpunkt: 265…275 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00094344 CAS-Nummer: 7159-36-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...