5-Hydroxy-2-adamantanon

Lieferant: Apollo Scientific

Synonyme: Idramantone, 5-Hydroxyadamantan-2-one

OR60074-1G
APOSOR60074-1GEA 21.9 EUR
APOSOR60074-1G
5-Hydroxy-2-adamantanon
5-Hydroxy-2-adamantanon
Formel: C₁₀H₁₄O₂
Molecular Weight: 166,22 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00192211
CAS-Nummer: 20098-14-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR