2-(2-Amino-1-hydroxyethyl)pyridine

Lieferant: Apollo Scientific
Achtung

OR60079-500MG
APOSOR60079-500MGEA 415 EUR
APOSOR60079-500MG
2-(2-Amino-1-hydroxyethyl)pyridine
2-(2-Amino-1-hydroxyethyl)pyridine
Formel: C₇H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD03840171
CAS-Nummer: 89943-14-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR