4-Chloro-6-methylpyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
OR60081-1G
APOSOR60081-1GEA
29
EUR
APOSOR60081-1G
4-Chloro-6-methylpyrimidine
4-Chloro-6-methylpyrimidine
Formel:
C₅H₅ClN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02322991 CAS-Nummer: 3435-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...