4-Amino-2-chlorphenol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chlor-4-hydroxyanilin

OR60093-100G
APOSOR60093-100GEA 159 EUR
APOSOR60093-100G
4-Amino-2-chlorphenol 97%
4-Amino-2-chlorphenol
Formel: C₆H₆ClNO
Molecular Weight: 143.57 g/mol
Schmelzpunkt: 150…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00128891
CAS-Nummer: 3964-52-1
EINECS: 202-458-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR