1-Benzyl-2-(methylthio)-1H-imidazole-5-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR60098-100MG
APOSOR60098-100MGEA
21.9
EUR
APOSOR60098-100MG
1-Benzyl-2-(methylthio)-1H-imidazole-5-carboxylic acid
1-Benzyl-2-(methylthio)-1H-imidazole-5-carboxylic acid
Formel:
C₁₂H₁₂N₂O₂S Molecular Weight: 248,301 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04974044 CAS-Nummer: 403479-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...