2,6-Dichlor-4-methylpyridin

Lieferant: Apollo Scientific

OR60099-1G
APOSOR60099-1GEA 21.9 EUR
APOSOR60099-1G
2,6-Dichlor-4-methylpyridin
2,6-Dichloro-4-methylpyridine
Formel: C₆H₅Cl₂N
Molecular Weight: 162,02 g/mol
MDL: MFCD09264302
CAS-Nummer: 39621-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR