6-Chloro-3,4-pyridinediamine
Lieferant: Apollo Scientific
|
Achtung
|
OR60103-250MG
APOSOR60103-250MGEA
29
EUR
APOSOR60103-250MG
6-Chloro-3,4-pyridinediamine
6-Chloro-3,4-pyridinediamine
Formel:
C₅H₆ClN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037269 CAS-Nummer: 89182-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...