1H-Indazol-5-boronsäure

Lieferant: Apollo Scientific
Achtung

OR60111-1G OR60111-250MG
APOSOR60111-1GEA 56.7 EUR
APOSOR60111-1G APOSOR60111-250MG
1H-Indazol-5-boronsäure
1H-Indazole-5-boronic acid
Formel: C₇H₇BN₂O₂
Molecular Weight: 161.9537 g/mol
Schmelzpunkt: 175…180 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06739151
CAS-Nummer: 338454-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR