5-Brom-3-iod-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
OR60116-1G
OR60116-250MG
OR60116-5G
APOSOR60116-1GEA
75.7
EUR
APOSOR60116-1G
APOSOR60116-250MG
APOSOR60116-5G
5-Brom-3-iod-1H-indazol
5-Bromo-3-iodo-1H-indazole
Formel:
C₇H₄BrIN₂ Molecular Weight: 322.9300 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07781637 CAS-Nummer: 459133-66-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...