Methyl-3-iod-1H-indazol-6-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-iodo-1H-indazole-6-carboxylate

OR60120-1G
APOSOR60120-1GEA 21.9 EUR
APOSOR60120-1G
Methyl-3-iod-1H-indazol-6-carboxylat
Methyl-3-iod-1H-indazol-6-carboxylat
Formel: C₉H₇IN₂O₂
Molecular Weight: 302,0686 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781616
CAS-Nummer: 885518-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR