6-Amino-4-methylnicotinonitrile
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
6-Amino-4-methylpyridine-3-carbonitrile, 2-Amino-5-cyano-4-picoline
OR60121-250MG
APOSOR60121-250MGEA
21.9
EUR
APOSOR60121-250MG
6-Amino-4-methylnicotinonitrile
6-Amino-4-methylnicotinonitrile
Formel:
C₇H₇N₃ Molecular Weight: 133,151 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09839255 CAS-Nummer: 179555-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...