2-Amino-4-(3-nitrophenyl)pyrimidine 97%

Lieferant: Apollo Scientific

Synonyme: 4-(3-Nitrophenyl)pyrimidin-2-amine

OR60125-250MG
APOSOR60125-250MGEA 85.7 EUR
APOSOR60125-250MG
2-Amino-4-(3-nitrophenyl)pyrimidine 97%
2-Amino-4-(3-nitrophenyl)pyrimidine
Formel: C₁₀H₈N₄O₂
Molecular Weight: 216,196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665910
CAS-Nummer: 240136-69-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR