(5-Chloro-2-pyridyl)methanamine dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Aminomethyl)-5-chloropyridine dihydrochloride

OR60126-250MG
APOSOR60126-250MGEA 21.9 EUR
APOSOR60126-250MG
(5-Chloro-2-pyridyl)methanamine dihydrochloride
(5-Chloro-2-pyridyl)methanamine dihydrochloride
Formel: C₆H₉Cl₃N₂
Molecular Weight: 215,51 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16872047
CAS-Nummer: 1257535-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR