1-Hydroxy-2-naphthoesäure
Lieferant: Apollo Scientific
|
Gefahr
|
OR60129-100G
OR60129-250G
OR60129-5G
OR60129-500G
APOSOR60129-100GEA
0
EUR
APOSOR60129-100G
APOSOR60129-250G
APOSOR60129-5G
APOSOR60129-500G
1-Hydroxy-2-naphthoesäure
1-Hydroxy-2-naphthoesäure
Formel:
C₁₁H₈O₃ Flammpunkt: 150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003960 CAS-Nummer: 86-48-6 EINECS: 201-674-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...