Ethyl 1,3,5-trimethyl-1H-pyrazole-4-carboxylate
Lieferant: Apollo Scientific
Synonyme:
4-(Ethoxycarbonyl)-1,3,5-trimethyl-1H-pyrazole
OR60138-1G
APOSOR60138-1GEA
72.5
EUR
APOSOR60138-1G
Ethyl 1,3,5-trimethyl-1H-pyrazole-4-carboxylate
Ethyl 1,3,5-trimethyl-1H-pyrazole-4-carboxylate
Formel:
C₉H₁₄N₂O₂ Molecular Weight: 182,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00159644 CAS-Nummer: 56079-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...