(±)-δ-Decalacton 95+%
Lieferant: Apollo Scientific
Synonyme:
(±)-5-Hydroxydecansäurelacton, (±)-δ-Dekalacton, (±)-Decan-5-olid, (±)-Tetrahydro-6-pentyl-2H-pyran-2-on
OR6014-100G
APOSOR6014-100GEA
32.2
EUR
APOSOR6014-100G
(±)-δ-Decalacton 95+%
(±)-δ-Decalacton
Formel:
C₁₀H₁₈O₂ Molecular Weight: 170.25 g/mol Siedepunkt: 117…120 °C/0.02mmHg Dichte: 0.954 g/cm³ (25 °C) Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006649 CAS-Nummer: 705-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...