2-(Aminomethyl)-5-bromo-4-methylpyridine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Bromo-4-methylpyridin-2-yl)methanamine hydrochloride, 5-Bromo-4-methylpicolinylamine hydrochloride, (5-Bromo-4-methylpyridin-2-yl)methylamine hydrochloride

OR60141-250MG
APOSOR60141-250MGEA 151 EUR
APOSOR60141-250MG
2-(Aminomethyl)-5-bromo-4-methylpyridine hydrochloride
2-(Aminomethyl)-5-bromo-4-methylpyridine hydrochloride
Formel: C₇H₉BrN₂·ClH
Molecular Weight: 237,525 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17214201
CAS-Nummer: 1257535-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR