5-Chloro-3-methylpyridine-2-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-3-methylpicolinonitrile, 5-Chloro-2-cyano-3-methylpyridine

OR60143-1G
APOSOR60143-1GEA 43.5 EUR
APOSOR60143-1G
5-Chloro-3-methylpyridine-2-carbonitrile
5-Chloro-3-methylpyridine-2-carbonitrile
Formel: C₇H₅ClN₂
Molecular Weight: 152,581 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17214209
CAS-Nummer: 156072-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR