6-Chloro-4-methylnicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-4-methyl-3-pyridinecarboxylic acid, 6-Chloro-4-methylpyridine-3-carboxylic acid, 5-Carboxy-2-chloro-4-methylpyridine, 2-Chloro-4-methylpyridine-5-carboxylic acid

OR60150-1G
APOSOR60150-1GEA 60.9 EUR
APOSOR60150-1G
6-Chloro-4-methylnicotinic acid
6-Chloro-4-methylnicotinic acid
Formel: C₇H₆ClNO₂
Molecular Weight: 171,581 g/mol
Flammpunkt: >100 °C
Lagertemperatur: Kühlschrank
MDL: MFCD12755871
CAS-Nummer: 503555-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR