5-Bromo-2-chloro-3-methoxypyridine

Lieferant: Apollo Scientific
Gefahr

OR60155-5G OR60155-10G
APOSOR60155-5GEA 277 EUR
APOSOR60155-5G APOSOR60155-10G
5-Bromo-2-chloro-3-methoxypyridine
5-Bromo-2-chloro-3-methoxypyridine
Formel: C₆H₅BrClNO
Molecular Weight: 222.47 g/mol
Schmelzpunkt: 68…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08234976
CAS-Nummer: 286947-03-3
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR