4-Isoxazolcarbonsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Isoxazole-4-carboxylic acid
OR60164-5G
APOSOR60164-5GEA
163
EUR
APOSOR60164-5G
4-Isoxazolcarbonsäure 98%
4-Isoxazolecarboxylic acid
Formel:
C₄H₃NO₃ Molecular Weight: 113.0715 g/mol Schmelzpunkt: 121…123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06738806 CAS-Nummer: 6436-62-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...