2-Ethyl-5-nitrobenzoxazol

Lieferant: Apollo Scientific
Achtung

OR60171-1G
APOSOR60171-1GEA 53.8 EUR
APOSOR60171-1G
2-Ethyl-5-nitrobenzoxazol
2-Ethyl-5-nitrobenzoxazole
Formel: C₉H₈N₂O₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 204771-74-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR