2-Propyl-5-nitro-1,3-benzoxazole

Lieferant: Apollo Scientific

Synonyme: 5-Nitro-2-propyl-1,3-benzoxazole

OR60172-1G
APOSOR60172-1GEA 139 EUR
APOSOR60172-1G
2-Propyl-5-nitro-1,3-benzoxazole
2-Propyl-5-nitro-1,3-benzoxazole
Formel: C₁₀H₁₀N₂O₃
Molecular Weight: 206,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06496176
CAS-Nummer: 861211-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR