Piperazine-1-carboximidamide

Lieferant: Apollo Scientific
Achtung

OR60177-1G
APOSOR60177-1GEA 0 EUR
APOSOR60177-1G
Piperazine-1-carboximidamide
Piperazine-1-carboximidamide
Formel: C₅H₁₂N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD04116543
CAS-Nummer: 45695-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR