6,7-Dihydrocyclopenta[b]pyridin-5-one

Lieferant: Apollo Scientific
Achtung

OR60178-1G
APOSOR60178-1GEA 52.2 EUR
APOSOR60178-1G
6,7-Dihydrocyclopenta[b]pyridin-5-one
6,7-Dihydrocyclopenta[b]pyridin-5-one
Formel: C₈H₇NO
Molecular Weight: 133,15 g/mol
Schmelzpunkt: 64…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07371500
CAS-Nummer: 28566-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR