4-Bromo-1H-pyrazole-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR60183-1G
OR60183-250MG
APOSOR60183-1GEA
21.9
EUR
APOSOR60183-1G
APOSOR60183-250MG
4-Bromo-1H-pyrazole-3-carboxylic acid
4-Bromo-1H-pyrazole-3-carboxylic acid
Formel:
C₄H₃BrN₂O₂ Molecular Weight: 190.9828 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD01459882 CAS-Nummer: 13745-17-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...